Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Nickel-Chromium Crucible

Catalog No. 13812121 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Outer:
44 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer
13-812-121 44 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-812-121
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fits Nickel-Chromium Crucible (Catalogue number: 13812120)

Fisherbrand™ Nicrhome Crucible Cover

  • Cover for 13-812-120 Crucible
  • 1D × 0.43 in.H (26 × 11mm)

Specifications

Material Nickel Chromium
For Use With (Application) For nickel chromium crucible
Diameter (Metric) Outer 44 mm
Height (Metric) 11mm
Product Type Crucible Cover
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.