Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Addressograph - Patient ID Labels
GREENER_CHOICESDP

Smudge-proof, for use on clean, flat surfaces.

$83.90 - $108.60

Specifications

Color White
Format Label
Product Type Patient ID Label
For Use With (Equipment) Addressograph and other patient name plate imprinters
Quantity 1000/Roll
View More Specs

Products 2
Catalog Number Size (English) Size (Metric) Price Quantity & Availability  
Catalog Number Size (English) Size (Metric) Price Quantity & Availability  
15920
GREENER_CHOICESDP
View Documents
1 x 3 in. 2.5 x 7.6 cm
Case of 1000 for $108.60
Only null left
Add to Cart
 
15921
GREENER_CHOICESDP
View Documents
1.5 x 3.5 in. 3.81 x 8.89 cm
Case of 1000 for $83.90
Only null left
Edge
Add to Cart
 
Description

Description

  • White labels for use with Addressograph and other patient name plate imprinters
  • Peel-off labels are die-cut on paper backing with perforations between each label
  • Removable

Order Info

1000 labels per roll.

Specifications

Specifications

White
Patient ID Label
1000/Roll
Label
Addressograph and other patient name plate imprinters
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.