Learn More
Fisherbrand™ Pellet Pestle™ Cordless Motor
SDP
Catalog No. 12141361 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Pellet Pestle Cordless Motor
Catalog No. Product Type
12-141-361 Pellet Pestle Cordless Motor
1 options
Catalog No. 12-141-361
Add to Cart
Add to Cart
Produces more complete disaggregation than manual mixing. The Pellet Pestle™ Cordless Motor can be used with disposable or reusable Pellet Pestle™ Mixers.
  • Use with either disposable or reusable Pellet Pestle™ Mixers
  • Comes with two “AA” batteries
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Pellet pestle mixer
For Use With (Application) Either disposable or reusable pellet pestle mixers, resuspend Protein and DNA pellets or grind soft tissues in microcentrifuge tubes
Includes 2 AA batteries
Material Other
Product Type Pellet Pestle Cordless Motor