Learn More
Description
- Patented double-clicking lock design
- 1st audible“click”ensures closure
- 2nd audible“click”achieves full 95kPa certification
- Available in two convenient sizes –20mL (10mL filled) neutral-buffered formalin;60mL (30mL filled) with wide mouth (53mm) opening neutral-buffered formalin
- Leak tested and independently 95kPa certified
- Avoid employee/patient exposure to hazardous chemicals
- Improve efficiency of specimen transport to the laboratory
- Economical bulk packaging with inner convenient transport packs help grow your Outreach Programs
Specifications
Specifications
| Material | Polypropylene |
| Sterility | Non-sterile |
| Capacity (Metric) | 60 mL, pre-filled 30 mL |
| Closure Material | Polyethylene |
| Closure Type | Permaloc™ Screw Cap |
| Color | Clear |
| Graduations | None |
| Height (Metric) | 6 cm |
| Length (Metric) | 5.1 cm |
| No. per Case | 324/Cs. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
