Learn More
Fisherbrand™ Photoionization Lamps
Catalog No. 14386113C Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
OI/Hewlett-Packard GC detector
Photovac/Thermo environmental
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-386-113C OI/Hewlett-Packard GC detector
14-386-113D Photovac/Thermo environmental
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-386-113C
Add to Cart
Add to Cart
Available in the DC style in the 10.0 to 10.6eV range commonly used for most analyses
  • Popular in many environmental and gas chromatographic detector applications
  • High energy is guaranteed through the use of ideally dimensioned windows unique to these lamps
  • Low transmission losses and resistance to long-term aging ensure high analytical sensitivity
  • Lifetime in excess of 3000 hours at ambient temperature can be expected with typical operation
  • Windows are MgF2
  • Krypton gas fill
Specifications
Gas Fill Krypton
For Use With (Equipment) OI/Hewlett-Packard GC detector
Output 10/10.6 eV
Product Type Photoionization Lamp