Learn More
Description
Provides necessary suction to fill standard glass and plastic pipets safely. Assorted colors, sizes and styles
- For use with pipets
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Black |
| For Use With | Pipette |
| Type | Bulb, with metal valve |
| Volume (Metric) | 75 mL |
| For Use With (Equipment) | Pipets |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.