Description
- One-piece molded construction with permanently molded-on graduations
- New resin formulation provides greater clarity
- Exposure to temperatures above 85°C (185°F) may alter accuracy
- Repeated autoclaving may alter accuracy
Specifications
Specifications
| Material | Polypropylene |
| Graduations | 2 mL |
| Product Type | Ungraduated Cylinder |