Learn More
Fisherbrand™ Polypropylene Transport Tubes
Catalog No. p-150487 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Nonsterile
Sterile
Capacity (Metric):
5 mL
10 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Capacity (Metric)
02-681-130 Nonsterile 5 mL
02-681-131 Nonsterile 10 mL
02-681-132 Sterile 5 mL
02-681-133 Sterile 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-681-130
  • Disposable
  • Frosted writing area
  • Molded-in graduation marks and sealing rings
TRUSTED_SUSTAINABILITY
Specifications
Type Nonsterile
Closure Type Blue Screw
Capacity (Metric) 5 mL
Description ATP, RNase/DNase, DNA, PCR inhibitor and Endotoxin Free (non-pyrogenic)
Material Polypropylene