Learn More
Fisherbrand™ Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
 View more versions of this product
Catalog No. FB965R
Add to cart
Description
Description
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
  • Glazed inside and out except for the outside bottom surface
  • With perforated handle
  • Strongly recommended that heating and cooling rates not exceed 200°C per hour and maximum use temperature of 1100°C
Specifications
Specifications
Crucible Cover
6g
With Perforated handle
30mm
Use with Fisherbrand high and wide form porcelain crucible
1150°C
1.75mm
Porcelain
26mm
Glazed
5mm