Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Porcelain Rose Crucible

Catalog No. FB960Z Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
FB960Z 75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FB960Z

missing translation for 'validMainframeMsg'

For use in very high temperatures

  • Unglazed
  • Without perforated cover or tube
  • Color: white
  • Capacity: 2.54 oz. (75mL)

Specifications

Material Porcelain
For Use With (Application) Used in very high temperature
Capacity (Metric) 75 mL
Product Type Rose Crucible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.