Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Premium Foam-Insulated Leather Drivers Gloves

Catalog No. 19013381 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-013381 X-Large
19-013379 Large
19-013374 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-013381

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Constructed from top grain cowhide leather with foam insulation to keep hands warm during cold applications.

  • Ideal for cold weather applications such as farming, construction, and refrigeration
  • Straight thumb and shirred elastic back
  • Red foam lining adds comfort and warmth
  • Easy size identification by color and cuff
  • Gunn cut

Specifications

Color Natural
Grade Premium
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size X-Large
Style Gunn
For Use With (Application) Farming, Construction, Machine Operation, and Refrigeration
Glove Cuff Style Open
Hem Color Blue
Liner Material Foam
Series 77-208
Length (English) 10.7 in.
Length (Metric) 27.3 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.