Learn More
Description
Includes
Fitted PTFE covers or tapered PTFE stoppers, except 14-385-906A and -902A, which come with glass cover plates
- Quartz - high UV transmission and low thermal expansion
- Glass - used in the visible range of the spectrum
- Low values of refractive index and dispertion
- Good resistance to chemicals
Specifications
Specifications
| Material | Suprasil Quartz |
| Includes | PTFE Cover |
| Quantity | Ea. |
| Path Length | 5 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.