Learn More
Fisherbrand™ Racer Series Top Fuel Eyewear
Catalog No. 190421006 Shop All Fisherbrand Products
Change view
Click to view available options
Frame Color:
Black
Blue
Lens Tint:
Clear
Dark Gray
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-042-1006 Black Dark Gray
19-042-1005 Black Clear
19-042-1008 Blue Clear
19-042-1009 Blue Dark Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-042-1006
This item is not returnable. View return policy
Exceptional protection and style at an affordable price
  • Features a single-lens design with a sporty, dual-lens look
  • Nose bridge and bayonet-style side arms grip for a comfortable, confident fit
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
Lens Tint Dark Gray
Frame Color Black
Lens Material Polycarbonate