Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Red-Spirit™ Fractional-Degree Thermometers

Catalog No. 1504113B Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
-1°C to +101°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
15-041-13B -1°C to +101°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-041-13B

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Partial-immersion thermometers

  • Very accurate temperature readings without the environmental hazards of mercury
  • Graduated in 0.1°C divisions
  • Thermometers with low ranges of -200°C to +30°C are filled with pentane; -100°C to +50°C with xylene mixture
  • Red liquid thermometers at other ranges are filled with kerosene

Certifications

NIST traceable, Accuracy to ANSI™/SAMA tolerances

Specifications

Length (English) Overall 23.8 in.
Length (Metric) Overall 610 mm
Max. Temperature (Metric) 101°C
Min. Temperature (Metric) -1°C
Temperature Range (Metric) -1°C to +101°C
Immersion Length 76 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.