Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MBP Rimmed Microcentrifuge Tube Plug Closures
GREENER_CHOICE

Catalog No. 02681450 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-681-450 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-681-450 Supplier Thermo Scientific™ Supplier No. 506FIS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polypropylene snap cap tubes with indented top

  • Plug Closures
  • Fit 1.5 and 2.0mL tubes
  • Color: white

TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Color White
For Use With (Equipment) 1.5 and 2 mL Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.