Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Serum Transport Tubes

Catalog No. 22038770 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22038770 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22038770 Supplier Fisherbrand™ Supplier No. M995200

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Polypropylene tubes

  • 10 mL screw top Polypropylene tubes
  • Freestanding with skirted round bottom
  • Graduations in 1 mL increments up to 8 mL
  • Non-Sterile
  • 1000 per case
Note: Caps are not included; sold separately (see S40200, S40201, S40202, S40203, and S40204).

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Capacity (Metric) 10 mL
Length (Metric) 86 mm
Material Polypropylene
Quantity 1000/Cs.
Sterility Non-sterile
Diameter (Metric) 15 mm
Closure Type Not Included
Product Type Transport Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.