Learn More
Description
Exhaust ports are on the top or rear mounted. Can easily accomodate Microwave Tissue Processor and its accessories. (Contact your local architect and/or mechanical contractor to determine what accessories will be required to comply with local building codes.)
Requires ducting to HVAC system by user
Top (-TV) or rear (-RV) 6 in. O.D. exhaust ports
- External dimensions: 36W x 26D x 28 in.H (91 x 66 x 71cm)
- Internal dimensions: 34 x 25 x 27 in. (86 x 64 x 69cm)
- Weight: 26 lb. (12kg)
- Accommodates the following number of reagent bowls when used with Microwave Tissue Processor and optional Reagent Bowl Shelf (22047211): 4 large, 4 med. or 4-6 small bowls
- External dimensions: 48W x 26D x 28 in.H (122 x 66 x 71cm)
- Internal dimensions: 46 x 25 x 27 in. (117 x 64 x 69cm)
- Weight: 32 lb. (15kg)
- Accommodates the following number of reagent bowls when used with Microwave Tissue Processor and optional Reagent Bowl Shelf (22047212): 8 large, 8 med. or 10 small bowls
Specifications
Specifications
| Depth (English) | 26 in. |
| Depth (English) Interior | 27 in. |
| Depth (Metric) | 66 cm |
| Depth (Metric) Interior | 64 cm |
| Exhaust | Rear Exhaust ports |
| Height (English) | 28 in. |
| Height (English) Interior | 27 in. |
| Height (Metric) | 71 cm |
| Weight (English) | 26 lb. |
| Weight (Metric) | 12 kg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
