Learn More
Fisherbrand™ Siteliner Safety Glasses, Blue frame
Catalog No. 191302097 Shop All Fisherbrand Products
Change view
Click to view available options
Frame Color:
Black
Blue
Red, White, Black
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color
19-130-2097 Red, White, Black
19-130-2095 Blue
19-130-2096 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-130-2097
This item is not returnable. View return policy
Unilens design offers wide field of vision and exceptional clarity
  • Lightweight, impact-resistant nylon frames
  • One-piece, clear hardcoat polycarbonate lens for maximum protection
  • Scratch resistant; absorb >99.9% of harmful UV light
  • Molded-in sideshields and browguards for added protection against UV, dust and flying particles
  • Adjustable temples and lens inclination
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating Hardcoat
Shape Rectangle
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
Lens Tint Clear
Product Type Safety Glasses
Frame Color Red, White, Black
Material Polycarbonate (Lens), Nylon (Frame)
Lens Material Polycarbonate