Description
Includes
Infectious Substance and Dry Ice Super Labels
- For shipping infectious substances or diagnostic specimens that need to be shipped on dry ice
- Durable, lightweight
- Holds approximately 22 lb. (10kg) of dry ice, keeping contents frozen for up to 76 hours
Specifications
Specifications
| Quantity | 10/Cs. |
| Applications | For shipping infectious substances or diagnostic specimens that need to be shipped on dry ice |
| Type | Replacement Outer Box |