Learn More
Fisherbrand™ Stainless-Steel Gas Line Low-Pressure Filter Holder
Catalog No. 0975313C Shop All Fisherbrand Products
Change view
Click to view available options
Gas Line Holder Ss 47mm
Catalog No.
09-753-13C Gas Line Holder Ss 47mm
1 options
Catalog No. 09-753-13C
Recommended for inline ultracleaning/sterilization of air, gases, compressed gases, and liquids at ≤50psi. Ideal for filtering pressure vessel gas inlets, gases from pressure pumps. (With gases containing moisture or water droplets, use 0.5μm pore size PTFE membranes) Feature threaded female locking nut—lets you change filters without removing holder from gas lines; can be used with flow in either direction. Two stainless-steel screens provide filter and back-pressure support; protect membrane.
Includes Type 316 stainless-steel inlet and outlet bodies; two support screens (back pressure and photo etched), and locking nut; under-drain screen; nickel-plated pipe plug; and one O-ring. Upstream and downstream adapters are available; sold separately (NC9286360). Autoclavable, with or without membrane in place.
†Autoclavable with or without membrane in place.
†At 10psi differential pressure.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Filtration Area 13.8 cm2
Max. Pressure 20 bar (290psi)
Type Gas Line Holder