Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Standard Disposable Transfer Pipettes, Nongraduated; Length: 5.875 in.; Capacity: 7.7 mL

Catalog No. 137117M Shop All Fisherbrand Products
Change view
Click to view available options
Nongraduated; Length: 5.875 in.; Capacity: 7.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
13-711-7M Nongraduated; Length: 5.875 in.; Capacity: 7.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-711-7M

missing translation for 'validMainframeMsg'

Single squeeze draws up to 3.2 mL

Unbreakable all-in-one pipettes eliminate the hazards of broken glass and exposure to infectious materials.

  • Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel
  • One-piece design molded from see-through, low-density polyethylene
  • Inert to biological fluids and most acids
  • Work well anywhere there is a need for quick, safe transfer of fluids
  • In four ready-to-use styles
  • One squeeze draws from 2.0 to 3.3 mL (depending on style used)
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)
TRUSTED_SUSTAINABILITY

Specifications

Bulb Draw (Metric) 3.2 mL
Material Polyethylene
Sterility Non-sterile
For Use With (Application) Research
Graduated No
Quantity 500/Pk.
Volume (Metric) 7.7 mL
Length (Metric) 15 cm
Type Transfer Pipet
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.