Learn More
Fisherbrand™ Stirring Paddles
Ideal for use in stirring small volumes or for chemical mixing in photographic work.
$138.46 - $417.96
Specifications
For Use With (Application) Stirring small volumes or for chemical mixing in photographic work
View More Specs
Products 3
Catalog Number Length (English) Handle Dimensions (L x W) Price Quantity & Availability  
Catalog Number Length (English) Handle Dimensions (L x W) Price Quantity & Availability  
14-243-5A
View Documents
20 in. Paddle: 2 x 3 in.
Case of 4 for $199.58
Only null left
Edge
Add to Cart
14-243-5C
View Documents
72 in. Paddle: 3 x 6 in.
Each for $138.46
Only null left
Add to Cart
14-243-5B
View Documents
36 in. Paddle: 3 x 6 in.
Case of 4 for $417.96
Only null left
Add to Cart
Description
Description
Choose from small or large size paddles.
  • Made from high-density polyethylene
  • Small paddle is ideal for use in stirring small volumes or for chemical mixing in photographic work
  • Large paddles are excellent for mixing liquids in large tanks or vats
Order Info
Small paddle handle dia. is 0.5 in. (1.3cm); large paddle handle dia. is 1 in. dia. (2.5cm). Additional stirring paddle sizes available. To order, contact your Fisher Scientific Customer Service Representative.
Specifications
Specifications
Stirring small volumes or for chemical mixing in photographic work
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.