Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Straight Medium Point Forceps with Guide Pin

Catalog No. 16100106 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
115 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
16-100-106 115 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-100-106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features a guide pin for perfect alignment

  • Straight serrated medium tip
  • Finger serrations for added control
  • Made of stainless steel
  • Length, 4.5 in., ; L: 4.5 in. (115mm); W: 0.32 in. (8mm); H: 0.06 in. (1.5mm)

Specifications

Product Type Forceps
Length (English) 4.5 in.
Length (Metric) 115 mm
Material Stainless Steel
For Use With (Application) Dissection, research, laboratories, small-parts handling
Includes Serrated Tip and Handles; Guide Pin
Tip Style Straight, Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.