Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Straight Strong Point Boley Style Forceps

Catalog No. 16100124 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
127 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
16-100-124 127 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-100-124
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flexible, general purpose forceps

  • Made of antimagnetic stainless steel
  • Length 5 in., L: 5 in. (127mm); W: 0.443 in. (11mm); H: 0.06 in. (1.5mm)

Specifications

Product Type Forceps
Length (English) 5 in.
Length (Metric) 127 mm
Material Stainless Steel
For Use With (Application) Dissection, research, laboratories, small-parts handling
Includes No Serrations
Tip Style Straight, Strong
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.