Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Superfrost™ Clipped Corner Slides

Catalog No. 12550016 Shop All Fisherbrand Products
Change view
Click to view available options
Clipped Corner; Size: 25 x 75mm, 1mm thick
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
12-550-016 Clipped Corner; Size: 25 x 75mm, 1mm thick
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-550-016
Add to Cart
Add to Cart

45° clipped corner slide offers exceptional cleanliness and resistance to sticking

  • Designed specifically for the Sysmex™ SP-100 slide prep unit
  • Tight tolerances for the length and width help to reduce accumulation of glass chips within the instrument
  • Corners are safer, less prone to chipping during handling
  • Draws specimens away from edges
  • Durable, opaque printed surface inert to common laboratory chemicals and reagents
  • Autoclavable
  • Size: 25 × 75 × 1mm

Specifications

Length (Metric) 25 mm
Frosted SuperFrost
Thickness (Metric) 1 mm
Width (Metric) 75 mm
Corner Style Clipped
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.