Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ SURE-TEK™ 2 Processing Embedding Cassettes Dispenser Packs
SDP

Catalog No. 22048141 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Aqua
Blue
Green
Lilac
Pink
White
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-048-141 White
22-048-142 Blue
22-048-143 Green
22-048-145 Pink
22-048-150 Lilac
22-048-149 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-048-141
This item is not returnable. View return policy

Comes with attached lids

  • Supplied in Dispenser Packs
  • Available in White, Blue, Green, Grey, Pink, Peach, Yellow, Tan, Aqua and Lilac colors

Specifications

For Use With (Equipment) Sure-tek™ 2 Cassette
Includes Attached lid
Type Processing and Embedding Cassette
Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.