Learn More
Fisherbrand™ Traceable™ Auto Shut-Off Timer
Catalog No. 15079550 Shop All Fisherbrand Products
Change view
Click to view available options
Battery Life:
12 Months
Catalog No. Battery Life
15-079-550 12 Months
1 options
Catalog No. 15-079-550
Add to Cart
Add to Cart
Timer automatically shuts off after 2 minutes of inactivity to conserve battery power.
With a modern design, polished metal case and rubberized backing, this Traceable Timer is an attractive addition to any lab. Use fast-forward function to quickly set the countdown time. Set any countdown time from 99 minutes, 59 seconds to 1 second with an accuracy of 0.01%. Additional features include Automatic-Bounceback™ Memory for repetitive timing, stopwatch, and non-skid backing. Large sized keys and 1-inch sized display make it easy to set time and read from across the lab. Small size, 2-5/8 x 2-1/2 x 11/16 inches, allows the timer to easily be carried on hand or be moved to a variety of locations as needed.
Specifications
Type Timer, Single Channel
Display Type Digital
Calibration Calibrated
Alarm Types Audible
Software Type TraceableLIVE
Battery Life 12 Months
Includes Individually serial-numbered Traceable Certificate, magnetic back, easel stand and battery
Channels 1
Certifications/Compliance NIST, ISO 17025, A2LA
Accuracy 0.01%
Range 99 minutes, 59 seconds
Resolution 1
Dimensions (L x W x H) 2 5/8 x 11/16 x 2 1/2 in.