Learn More
Description
- For 10 and 12mL serum transport tubes
Compliance
95kPa compliant to DOT and IATA standards when combined with Fisherbrand threaded transport tubes
Specifications
Specifications
| Certifications/Compliance | DOT, IATA |
| Product Type | Screw Cap |
| Material | Polyethylene |
| Color | Blue |
| For Use With (Equipment) | 10 and 12 mL Serum Transport Tubes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
