Learn More
Fisherbrand™ Two-Hole Rubber Stoppers
SDP
Assorted sizes 00 to 15
 View more versions of this product
Catalog No. 14-140F
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Assorted sizes 00 to 15
  • Hole size: 3 to 5mm
Specifications
Specifications
Tapered Stopper (2 holes)
2 cm
4
2
General purpose use
Natural Rubber
2.6 cm
2.5 cm
4
5mm
0.5cm
Black
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.