Learn More
Fisherbrand™ Immersion Baskets for Ultrasonic Cleaners
SDP
This immersion basket is designed specifically for the Fisherbrand™ 112xx Series Dual Frequency Ultrasonic Cleaners. The fine mesh makes this immersion basket ideal for small items. The extra-long handle makes handling safe and easy.
 View more versions of this product
Catalog No. 2070380000
Description
Description
  • Fine mesh for small part security
  • Available in polypropylene or stainless steel
  • Extra-long handle for ease-of-use
Specifications
Specifications
Immersion Basket
FB11211 Ultrasonic Cleaning Unit
Polypropylene
74 x 50 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.