Learn More
Fisherbrand™ Immersion Baskets for Ultrasonic Cleaners
This immersion basket is designed specifically for the Fisherbrand™ 112xx Series Dual Frequency Ultrasonic Cleaners. The fine mesh makes this immersion basket ideal for small items. The extra-long handle makes handling safe and easy.
Description
- Fine mesh for small part security
- Available in polypropylene or stainless steel
- Extra-long handle for ease-of-use
Specifications
Immersion Basket | |
Stainless Steel | |
59 x 60 mm |
FB11211 Ultrasonic Cleaning Unit | |
Round |