Learn More
Fisherbrand™ Unisette™ Tissue Processing/Embedding Cassettes
Catalog No. 22038118 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-118 Yellow
22-038-124 Orange
22-038-117 Green
22-038-119 Blue
22-038-115 White
22-038-125 Aqua
22-038-122 Gray
22-038-123 Lilac
22-038-120 Peach
22-038-116 Pink
22-038-121 Tan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 22-038-118
Snap-latch and hinge-lock design prevent early separation of base and lid and allows for one-handed operation
  • Molded from a special high-density polymer that is totally resistant to the chemical action of histological solvents
  • Specimens stay safely submerged in liquid
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • Lids can be opened and closed as often as necessary, and always relock securely
  • Large labeling areas located on three sides of cassette
  • Anterior writing area at a 35-degree angle
Specifications
Color Yellow