Description
- Molded from a special high-density polymer that is totally resistant to the chemical action of histological solvents
- Specimens stay safely submerged in liquid
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- One-piece integral lid eliminates the need for separate steel lids
- Lids can be opened and closed as often as necessary, and always relock securely
- Large labeling areas located on three sides of cassette
- Anterior writing area at a 35-degree angle
Specifications
Specifications
| Color | Gray |