Description
- 18 gauge stainless-steel polished to satin finish
- Knocked down (KD) type shelf
- Die stamped and die formed shelf and brackets
- Furnished with 1.5 in. sanitary rolled rim with 1.25 in. turn-up edge on sides and rear
Specifications
Specifications
| Product Type | Wall Shelf |
| For Use With (Application) | Convenient storage space for spices, timers etc |
| Width (English) Shelf | 11.125 in. |