Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Bacteriology Culture Collection and Transport System

Catalog No. 1490724 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 Pk.
100 Cs.
50 Pk.
Media Type:
Amies Charcoal
Amies Charcoal Gel
Amies Clear Gel
Liquid Amies
Liquid Stuart
Modified Cary-Blair
Multi-Collection
Swab Configuration:
Dual Applicator
Single Applicator
Shaft Material:
Plastic
Straight Wire
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type Swab Configuration Shaft Material
14-907-24 50 Pk. Liquid Amies Dual Applicator Plastic
14-907-12 50 Pk. Amies Clear Gel Single Applicator Plastic
14-907-13 50 Pk. Amies Clear Gel Dual Applicator Plastic
14-907-17 50 Pk. Amies Charcoal Gel Dual Applicator Plastic
14-907-14 50 Pk. Amies Clear Gel Single Applicator Straight Wire
14-907-16 50 Pk. Amies Charcoal Single Applicator Plastic
14-907-18 50 Pk. Amies Charcoal Single Applicator Straight Wire
14-907-23 50 Pk. Liquid Amies Single Applicator Plastic
14-907-25 50 Pk. Liquid Amies Single Applicator Straight Wire
14-907-20 50 Pk. Liquid Stuart Single Applicator Plastic
14-907-21 50 Pk. Liquid Stuart Dual Applicator Plastic
14-907-22 50 Pk. Liquid Stuart Single Applicator Straight Wire
14-907-26 10 Pk. Modified Cary-Blair Dual Applicator Plastic
14-907-27 100 Cs. Multi-Collection Single Applicator Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 14-907-24
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the collection and/or transportation of a wide range of bacteria, viruses and facultative microorganisms. All transport swabs have a rayon tip. Shaft types include plastic, straight wire or flexible wire. Plastic-shaft tips are available with single or double swab.

Amies Clear provides excellent survival for many bacteria, including anaerobes, and is compatible with gram stain. Modified Amies Charcoal enhances survival of fastidious organisms including Neisseria gonorrhoeae by neutralizing any toxins present in the specimen. Both Amies Clear and Amies Charcoal provide excellent environments for the survival of anaerobes by completely immersing swabs into a gel with sodium thioglycollate.  Modified Liquid Stuarts provides excellent recovery in routine bacteriology and are compatible with gram stain and many rapid screening methods.

 

  • Lot number and expiration date printed on tube labels
  • Swabs are tipped with inert, nontoxic rayon
  • Color-coded caps
  • Gamma radiation sterilized

Multicollection and Transport Swabs
  • Complete collection and transportation system for viruses, chlamydia, mycoplasma and ureaplasma
  • Glass beads provide extra surface area for cell adhesion and facilitate spreading of the specimen in subsequent culturing
  • Scored Dacron™ (polyester)-tipped swab breaks off cleanly at the right length inside the tube
  • Cap automatically engages swab shaft on closing and holds it tightly on reopening
  • Approved for storage at room temperature for up to one year prior to use
Anaerobic Transport System
  • Self-contained, sterile, ready-to-use system
  • Includes a glass tube containing a semisolid, prereduced transport medium
  • Medium indicator turns lavender if the anaerobic system has been compromised
  • Cap contains a pierceable, nonlatex stopper for injecting fluids
  • Scored swab breaks easily to fit in the tube

 

Specifications

Product Type Transport Swab
Format Swab
Media Type Liquid Amies
Sterility Sterile
Swab Configuration Dual Applicator
Closure Color Red
For Use With (Application) For the collection and/or transportation of a wide range of bacteria, viruses and facultative microorganisms
Includes Sponge insert at bottom of tube
Quantity 50 Pk.
Shaft Material Plastic
Tip Material Rayon
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.