Learn More
Biolegend FITC anti-Asialo-GM1
Supplier:  Biolegend 146006
Encompass_Preferred
FITC anti-Asialo-GM1 [Poly21460]; Isotype: Rabbit Polyclonal IgG; Reactivity: Mouse, Rat; Apps: FC; Size: 100 ug
Catalog No. 50-207-1680
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.