Learn More
Cytek Biosciences Inc Antibodies FITC Anti-Human CD5 (L17F12) ..... Cat. No. 35-0058-T500 ..... Size 500 tests
Supplier:  Cytek Biosciences Inc 350058T500
Encompass_Preferred
Antibodies FITC Anti-Human CD5 (L17F12) ..... Cat. No. 35-0058-T500 ..... Size 500 testsThe L17F12 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is a member of the scavenger receptor superfamily and is present on approximately 70% of normal peripheral blood lymphocytes. CD5 is involved in modulating antigen receptor signaling in both T and B cells and acts through binding of CD72, its receptor expressed on B cells.
Catalog No. 50-210-3213
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.