Learn More
Biolegend FITC anti-mouse H-2Kk
Supplier:  Biolegend 114905
Encompass_Preferred
FITC anti-mouse H-2Kk [36-7-5]; Isotype: Mouse (A.TL) IgG2a, κ; Reactivity: Mouse k haplotype, Weakly cross-reacts with SJL/Hsd mice splenocytes; Apps: FC; Size: 50 ug
Catalog No. 50-163-515
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.