Learn More
Abcam FITC Conjugation Kit, Lightning-Link™, 3 x 10 μg
Description
- Rapid Conjugation: FITC labeling in under 4 hours with 30 seconds of hands-on time.
- High Efficiency: Ensures 100% antibody recovery.
- Versatility: Suitable for conjugating antibodies, proteins, and peptides.
- Applications: Labeled products are ready for use in Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Format | Kit |
| Shipping Condition | Ambient |
| Product Type | Protein Labeling Reagent |
| Content And Storage | Store at -20°C. |
| Target Molecule | Antibodies, Proteins, Peptides |
| Detection Method | Fluorescence |
| Label or Dye | FITC |
| Quantity | 3 x 10 μg |
| For Use With (Application) | Western Blotting, ELISA, Immunohistochemistry |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.