Learn More
Abcam FITC Conjugation Kit, Lightning-Link™, 3 x 100 μg
Description
- Rapid Conjugation: FITC labeling in under 4 hours with 30 seconds hands-on time.
- High Efficiency: 100% antibody recovery.
- Versatility: Suitable for conjugating antibodies, proteins, and peptides.
- Applications: Can be used in Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Antibodies, Proteins, Peptides |
| Shipping Condition | Ambient |
| Label or Dye | FITC |
| Product Type | Protein Labeling Reagent |
| Content And Storage | Store at -20°C. |
| Format | Kit |
| Detection Method | Fluorescence |
| Quantity | 3 x 100 μg |
| For Use With (Application) | Western Blotting, ELISA, Immunohistochemistry |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.