Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ FITC-geldanamycin
SDP

Catalog No. NB61081561 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB61081561 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB61081561 Supplier R&D Systems™ Supplier No. NB61081561
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

Specifications

Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Product Line Novus
Quantity 1 mg
For Use With (Application) Purple Solid. Soluble in DMSO (>10 mg/mL).
Target FITC-geldanamycin
Type Geldanamycin
Label or Dye FITC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.