Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FITC Inhibitor
SDP

Catalog No. NB5440 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB5440 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB5440 Supplier Novus Biologicals™ Supplier No. NB5440

Low molecular weight, organic compound that may help regulate biological process.

Biological Activity: Fluorescent reagent for the labeling of amines. Excitation maximum: 495nm; emission maximum approx 525nm.

Storage: Store at 4°C.

Note: Inhibitors are guranteed for 1 year from date of receipt.

Specifications

Description CAS: 3326-32-7
Formulation C21H11NO5S
For Use With (Application) Staining solution for proteins
Quantity 50 mg
Product Type Staining solution
Molecular Weight (g/mol) 389.38g/mol
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Content And Storage Room Temperature
Color Orange

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.