Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human CD7 Protein, Fc Tag
Supplier:  ACROBiosystems CD7HF258200UG
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human CD7, Fc Tag (CD7-HF258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa.
Catalog No. 50-210-6880
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.