Learn More
Medchemexpress LLC FITC-labeled ODN 1018 sodium | 98.7% | 8,171.00 Da | 10 MG

Supplier: Medchemexpress LLC HY150724B10MG
FITC-labeled ODN 1018 sodium is an FITC-conjugated CpG oligodeoxynucleotide that functions as a Toll-like receptor 9 (TLR9) agonist. Supplied as the sodium salt, it is used to evaluate CpG ODN cellular uptake and localization by confocal laser-scanning microscopy or flow cytometry. For research use only.
- FITC label with excitation 495 nm and emission 520 nm.
- CpG oligodeoxynucleotide sequence: d(P-thio)(T-G-A-C-T-G-T-G-A-A-C-G-T-T-C-G-A-G-A-T-G-A).
- Molecular weight approximately 8,171 Da.
- Purity 98.7% as supplied.
- Sodium salt form.
- Available in 1 mg, 5 mg, and 10 mg sizes; larger sizes available by quote.
- Intended for research use only; not for human or clinical use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.