Learn More
Invitrogen™ FITC Polyclonal Antibody, HRP

Description
This product has an Enzyme: Protein (Molar Ratio) of >1.5 HRP:IgG. Rabbit anti FITC antibody recognizes fluorescein and does not react with immunoglobulins from any species.
Specifications
Specifications
| Antigen | FITC |
| Applications | ELISA, Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 1% BSA and 0.0095% Methylisothiazolone |
| Gene Alias | Fluorescein |
| Host Species | Rabbit |
| Immunogen | FITC Isomer I. |
| Purification Method | Affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.