Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FITC-Val-Ala-Asp(OMe)-FMK

Catalog No. MP03FK03205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
3 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP03FK03205 5 mg
MP03FK03203 3 mg
MP03FK03201 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP03FK03205 Supplier MP Biomedicals™ Supplier No. 03FK03205
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FITC-Val-Ala-Asp(OMe)-FMK, Yellow Solid

FITC-Val-Ala-Asp(OMe)-FMK is an inhibitor for General Caspases.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Peptide content: 77.6
Color Yellow
Purity >95% by TLC
For Use With (Application) Cell Viability Analysis
Content And Storage Keep cool and dry
Quantity 5 mg
Product Type Proteins, Enzymes & Peptides
Molecular Weight (g/mol) 722.7
Form Solid
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.