Learn More
S-Curve™ Five Compartment Glove Dispenser with Single Lid, Front Access
Catalog No. 19999433 Shop All S-Curve Technologies Products
Change view
Click to view available options
Material:
Acrylic
PETG
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-999-433 Acrylic
19-999-438 PETG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-999-433 Supplier S-Curve™ Supplier No. GD510
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Dispenser features five fully enclosed compartments and a single, sloped lid to maintain HEPA flow. Front access holes allow for easy product dispensing.
  • Five individual compartments with a single, sloped lid to maintain HEPA flow
  • Each compartment features a front access hole for product dispensing
  • Can be mounted to the wall with heavy-duty brackets (included)
  • Full-length hinged lid increases durability
  • Flame-treated edges for a smooth finish
Specifications
Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 12 x 26 x 12 in.
Holds Gloves
Material Acrylic
Mounting Keyhole Slots
No. of Compartments 5
Product Style Top Loading
Includes Heavy-duty wall mounting bracket
Thickness (English) 0.25 in.