Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Fixable Viability Dye, 665/685 nm, Cell-based, 200 Tests

Catalog No. 502699016
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-269-9016 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-269-9016 Supplier Abcam Supplier No. AB314684200TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labeling in under 4 hours with 30 seconds of hands-on time and 100% antibody recovery.

  • Assay type: Cell-based
  • Applications: Fluorescence Microscopy, Flow Cyt
  • Detection method: Flow cytometry-fluorescent
  • Sample types: Adherent cells, Suspension cells
  • Results type: Quantitative
  • Reactive species: Mammals
  • Key benefits: Labeling in < 4 hrs with 30 secs hands-on time; 100% antibody recovery

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Reactivity Mammals
Shipping Condition Ambient
No. of Reactions 200 Tests
Label or Dye Fixable Viability Dye (665/685)
For Use With (Equipment) Flow Cytometer, Fluorescence Microscope
Format Kit
Content And Storage Store at -20°C.
Detection Method Fluorescence
Detection Location In-Cell Detection
Cross Reactivity Mammals
Sample Type Adherent cells, Suspension cells
Quantity 200 Tests
For Use With (Application) Fluorescence Microscopy, Flow Cytometry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.