Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Fixable Viability Dye, 665/685 nm, Cell-Based, 50 Tests
Description
- Assay Type: Cell-based
- Detection Method: Flow cytometry-fluorescent
- Applications: Fluorescence Microscopy, Flow Cyt
- Sample Types: Adherent cells, Suspension cells
- Reactive Species: Mammals
- Results Type: Quantitative
- Features: Labeling in < 4 hrs with 30 secs hands-on time. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Viable Cells |
| Reactivity | Mammals |
| Label or Dye | Fixable Viability Dye |
| No. of Reactions | 50 Tests |
| For Use With (Equipment) | Flow Cytometer, Fluorescence Microscope |
| Content And Storage | Store at -20°C. |
| Format | Kit |
| Detection Location | In-Cell Detection |
| Detection Method | Fluorescence |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?