Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Fixable Viability Dye, 750/777 nm, 200 Tests

Catalog No. 502699020
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-269-9020 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-269-9020 Supplier Abcam Supplier No. AB314682200TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides rapid cell viability labeling with 100% antibody recovery.

  • Assay type: Cell-based
  • Applications: Fluorescence Microscopy, Flow Cyt
  • Sample types: Adherent cells, Suspension cells
  • Reactive species: Mammals
  • Results type: Quantitative
  • Protocol features: Labeling in < 4 hrs with 30 secs hands-on time; 100% antibody recovery

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Reactivity Mammals
For Use With (Equipment) Flow Cytometer, Fluorescence Microscope
No. of Reactions 200 Tests
Label or Dye Fixable Viability Dye
Format Kit
Content And Storage Store at -20°C.
Detection Method Fluorescence
Detection Location In-Cell Detection
Cross Reactivity Mammals
Sample Type Adherent cells, Suspension cells
Quantity 200 Tests
For Use With (Application) Fluorescence Microscopy, Flow Cytometry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.